Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRPB2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548365

Catalog No. PA548365

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in pre-mRNA splicing. This protein is associated with snRNP U2. It binds stem loop IV of U2 snRNA only in presence of the U2A ft. protein.
Specifications

Specifications

SNRPB2
Polyclonal
PBS with 0.09% sodium azide
P08579
U2 small nuclear ribonucleoprotein B′′, U2 snRNP B′′, SNRPB2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6629
Western Blot
Unconjugated
SNRPB2
Liquid
SNRPB2
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SNRPB2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit