Learn More
Selleck Chemical LLCSupplier Diversity Partner SNS-314 10mg 1057249-41-8  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S115410MG
SNS-314 is a potent and selective inhibitor of Aurora A, Aurora B and Aurora C with IC50 of 9 nM, 31 nM, and 3 nM, respectively. It is less potent to Trk A/B, Flt4, Fms, Axl, c-Raf and DDR2. Phase 1.
Catalog No. 50-716-9