Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNS 314 mesylate, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1146618-41-8
C19H19ClN6O4S3
527.029
FYCODPVDEFFWSR-UHFFFAOYSA-N
sns-314 mesylate,unii-kgw32fdy3u,kgw32fdy3u,n-3-chlorophenyl-n'-5-2-thieno 3,2-d pyrimidin-4-ylamino ethyl-2-thiazolyl urea methanesulfonate,n-3-chlorophenyl-n'-5-2-thieno 3,2-d pyrimidin-4-ylamino ethyl-2-thiazolyl urea methanesulfonate 1:1,urea, n-3-chlorophenyl-n'-5-2-thieno 3,2-d pyrimidin-4-ylamino ethyl-2-thiazolyl-, methanesulfonate 1:1,sns 314 mesylate,sns314, sns-314 mesylate,1-3-chlorophenyl-3-5-2-thieno 3,2-d pyrimidin-4-ylamino ethyl thiazol-2-yl urea methanesulfonate
24995523
1-(3-chlorophenyl)-3-[5-[2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl]-1,3-thiazol-2-yl]urea;methanesulfonic acid
CS(=O)(=O)O.C1=CC(=CC(=C1)Cl)NC(=O)NC2=NC=C(S2)CCNC3=NC=NC4=C3SC=C4
This item is not returnable. View return policy
1146618-41-8
C19H19ClN6O4S3
527.029
FYCODPVDEFFWSR-UHFFFAOYSA-N
sns-314 mesylate,unii-kgw32fdy3u,kgw32fdy3u,n-3-chlorophenyl-n'-5-2-thieno 3,2-d pyrimidin-4-ylamino ethyl-2-thiazolyl urea methanesulfonate,n-3-chlorophenyl-n'-5-2-thieno 3,2-d pyrimidin-4-ylamino ethyl-2-thiazolyl urea methanesulfonate 1:1,urea, n-3-chlorophenyl-n'-5-2-thieno 3,2-d pyrimidin-4-ylamino ethyl-2-thiazolyl-, methanesulfonate 1:1,sns 314 mesylate,sns314, sns-314 mesylate,1-3-chlorophenyl-3-5-2-thieno 3,2-d pyrimidin-4-ylamino ethyl thiazol-2-yl urea methanesulfonate
24995523
1-(3-chlorophenyl)-3-[5-[2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl]-1,3-thiazol-2-yl]urea;methanesulfonic acid
CS(=O)(=O)O.C1=CC(=CC(=C1)Cl)NC(=O)NC2=NC=C(S2)CCNC3=NC=NC4=C3SC=C4

Potent pan-Aurora kinase inhibitor

Potent, ATP-competitive Aurora kinase inhibitor (IC50 values are 3, 9 and 31 nM for Aurora C, A and B, respectively). Exhibits both in vitro and in vivo activity against human cancer cell lines and tumor xenograft models. Displays additive antiproliferative effects in combination with chemotherapeutics such as carboplatin (Cat. No. 2626), gemcitabine (Cat. No. 3259) and 5-fluorouracil (Cat. No. 3257).
Quantity 10mg
Formula Weight 527.04
Percent Purity >98%
Chemical Name or Material SNS 314 mesylate

Recommended Storage : Store at -20°C