Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntrophin alpha-1 Mouse anti-Human, Clone: OTI1E9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525324

Catalog No. PIMA525324


Add to cart

Description

Description

Dystrophin is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes.
Specifications

Specifications

Syntrophin alpha-1
Monoclonal
0.57 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q13424
LQT12, SNT1, TACIP1, dJ1187J4.5, 59 kDa dystrophin-associated protein A1 acidic component 1, acidic alpha 1 syntrophin, alpha-1-syntrophin, pro-TGF-alpha cytoplasmic domain-interacting protein 1, syntrophin-1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6640
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1E9
Unconjugated
SNTA1
Liquid
SNTA1
Full length human recombit protein of SNTA1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit