Learn More
Syntrophin alpha-1 Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI2E12, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525322
Catalog No. PIMA525322
Add to cart
Description
Description
Dystrophin is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes.
Specifications
Specifications
Syntrophin alpha-1
Monoclonal
0.86 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q13424
LQT12, SNT1, TACIP1, dJ1187J4.5
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
362242, 485837, 6640, 709824
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2E12
Unconjugated
SNTA1
Liquid
Snta1
Full length human recombit protein of SNTA1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate, Rat