Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNTB1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548888

Catalog No. PA548888

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Adapter protein that binds to and probably organizes the subcellular localization of a variety of membrane proteins. May link various receptors to the actin cytoskeleton and the dystrophin glycoprotein complex.
Specifications

Specifications

SNTB1
Polyclonal
PBS with 0.09% sodium azide
Q13884, Q99L88
Beta-1-syntrophin, 59 kDa dystrophin-associated protein A1 basic component 1, DAPA1B, BSYN2, Syntrophin-2, Tax interaction protein 43, TIP-43, SNTB1, SNT2B1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
20649, 6641
Western Blot
Unconjugated
SNTB1
Liquid
SNTB1
KLH conjugated synthetic peptide between 243-271 amino acids from the Central region of human SNTB1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit