Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SNUPN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530351
Description
Recommended positive controls: NT2D1, IMR32, U87-MG. Predicted reactivity: Mouse (95%), Rat (94%), Chimpanzee (99%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.
Specifications
SNUPN | |
Polyclonal | |
Unconjugated | |
SNUPN | |
0610031A09Rik; hypothetical protein LOC515588; KPNBL; RNA U transporter 1; RNA, U transporter 1; Rnut1; Snupn; Snupn1; snurportin 1; Snurportin1; snurportin-1; snurportin-1-like protein; SPN1; wu:fc41a03; zgc:77819 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10073 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O95149 | |
SNUPN | |
Recombinant fragment corresponding to a region within amino acids 1 and 249 of Human Snurportin 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction