Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNUPN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153581AP

Catalog No. 50-173-0175


Add to cart

Description

Description

This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.
Specifications

Specifications

SNUPN
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95149, Q68FP5, Q80W37
KPNBL, RNA U transporter 1, RNUT1, SNUPN, snurportin 1, Snurportin1, SPN1
Rabbit
IgG
150 μL
-20°C
Primary
10073, 316108, 66069
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SNUPN
Liquid
Snupn
SNUPN Fusion Protein Ag7604
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit