Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX1 Rabbit anti-Human, Unconjugated, Clone: JG40-06, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  Invitrogen MA534808

Catalog No. PIMA534808


Add to cart

Description

Description

Positive Control: Human skin tissue lysate, A549, SH-SY-5Y, SiHa, rat epididymis tissue, human colon tissue, human breast cancer tissue.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.
Specifications

Specifications

SNX1
JG40-06
Unconjugated
SNX1
Liquid
SNX1
Recombinant protein within human SNX1 aa 50-250.
Protein A
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Q13596
HsT17379; MGC8664; SNX1; SNX1A; sorting nexin 1; Sorting nexin-1; VPS5
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles, store in dark
Primary
6642
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit