Learn More
SNX10 Mouse anti-Human, Clone: OTI3F1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808884
Catalog No. 50-166-9149
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Probable phosphoinositide-binding protein involved in protein sorting and membrane trafficking in endosomes. Plays a role in cilium biogenesis through regulation of the transport and the localization of proteins to the cilium. Required for the localization to the cilium of V-ATPase subunit ATP6V1D and ATP6V0D1, and RAB8A. Involved in osteoclast differentiation and therefore bone resorption.
Specifications
Specifications
SNX10
Monoclonal
Unconjugated
SNX10
Lyophilized
SNX10
Full length human recombit protein of human SNX10 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3F1
PBS with 8% trehalose and no preservative; pH 7.3
Q9Y5X0
OPTB8
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
29887