Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
SNX11 293T Cell Overexpression Lysate 2 (Denatured), Abnova
Manufacturer:  Abnova Corporation H00029916T02
Catalog No. 89-002-157
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_013323
6.8
Transient overexpression cell Lysate was tested with Anti-SNX11 antibody (H00029916-B02) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
SNX11 293T Cell Transient Overexpression Lysate(Denatured)
25.2
pCMV-SNX11 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.