Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX12 Mouse anti-Human, Clone: OTI5C3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene CF804754
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members. A similar protein in mouse may be involved in regulating the neurite outgrowth. Alternate splicing results in multiple transcript variants.Specifications
SNX12 | |
Monoclonal | |
Unconjugated | |
SNX12 | |
sorting nexin 12 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29934 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5C3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9UMY4 | |
SNX12 | |
Full length human recombit protein of human SNX12 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction