Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX12 Mouse anti-Human, Clone: OTI3A4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526685

Catalog No. PIMA526685


Add to cart

Description

Description

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members. A similar protein in mouse may be involved in regulating the neurite outgrowth. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

SNX12
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UMY4
MGC118982, MGC118983, SNX12, sorting nexin 12, Sorting nexin-12
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29934
Immunohistochemistry (Paraffin), Western Blot
OTI3A4
Unconjugated
SNX12
Liquid
SNX12
Full length human recombit protein of SNX12 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit