Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX14 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548638

Catalog No. PA548638

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays a role in maintaining normal neuronal excitability and synaptic transmission. May be involved in several stages of intracellular trafficking.
Specifications

Specifications

SNX14
Polyclonal
PBS with 0.09% sodium azide
Q8BHY8
Sorting nexin-14, SNX14
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
244962
Western Blot
Unconjugated
Snx14
Liquid
SNX14
KLH conjugated synthetic peptide between 580-608 amino acids from the Central region of human SNX14.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit