Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX15 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259133

Catalog No. NBP259133


Add to cart

Description

Description

SNX15 Polyclonal antibody specifically detects SNX15 in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

SNX15
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
clone iota unknown protein, HSAF001435, sorting nexin 15, sorting nexin-15
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human
Immunoprecipitation
Immunoprecipitation
Affinity Purified
SNX15
Maps to a region between 225 and 275
Immunogen affinity purified
RUO
Primary
29907
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only