Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX21 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548882

Catalog No. PA548882

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in several stages of intracellular trafficking.
Specifications

Specifications

SNX21
Polyclonal
PBS with 0.09% sodium azide
Q969T3
Sorting nexin-21, Sorting nexin L, SNX-L, SNX21, C20orf161, SNXL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
90203
Western Blot
Unconjugated
SNX21
Liquid
SNX21
KLH conjugated synthetic peptide between 210-236 amino acids from the Central region of human SNX21.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit