Learn More
SNX25 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00083891T01
Catalog No. 89-009-590
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
293T
ENST00000296779
6.8
Transient overexpression cell Lysate was tested with Anti-SNX25 antibody (H00083891-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
SNX25 293T Cell Transient Overexpression Lysate(Denatured)
56.36
pCMV-SNX25 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot