Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX25 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550770

Catalog No. PA550770


Add to cart

Description

Description

The antibody detects endogenous levels of total SNX25 protein.

SNX25 (sorting nexin 25), also known as SBBI31 or MSTP043, is an 840 amino acid protein suggested to function in several stages of intracellular trafficking. A member of the sorting nexin family, SNX25 contains one PX (phox homology) domain, an RGS domain and one PXA domain. May be involved in several stages of intracellular trafficking.
Specifications

Specifications

SNX25
Polyclonal
Unconjugated
SNX25
Liquid
SNX25
Synthetic peptide corresponding to a region derived from internal residues of human sorting nexin 25
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9H3E2
SBBI31, MSTP043
Rabbit
IgG
100 μL
-20°C
Primary
83891
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit