Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNX27 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA523025

Catalog No. PIPA523025


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with rat, bovine and zebrafish based on 100% sequence homology.

SNX27 encodes a member of the sorting nexin family, a diverse group of cytoplasmic and membrane-associated proteins involved in endocytosis of plasma membrane receptors and protein trafficking through these compartments. All members of this protein family contain a phosphoinositide binding domain (PX domain). A highly similar protein in mouse is responsible for the specific recruitment of an isoform of serotonin 5-hydroxytryptamine 4 receptor into early endosomes, suggesting the analogous role for the human protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNX27
Polyclonal
Unconjugated
SNX27
5730552M22Rik; ESTM45; ESTM47; KIAA0488; MAP-responsive gene protein; methamphetamine (MAP) responsive transcript 1; methamphetamine-responsive transcript 1; methamphetamine-responsive transcript 1 protein; Mrt1; MY014; PDZ protein Mrt1; PDZ-protein Mrt1; R75405; RP11-98D18.12-005; SNX27; sorting nexin family member 27; sorting nexin-27
Rabbit
Antigen Affinity Chromatography
RUO
260323, 76742, 81609
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 30% glycerol and 0.1% sodium azide
Q3UHD6, Q8K4V4, Q96L92
SNX27
Synthetic peptide made to an internal portion of the human protein (within residues 450-500).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.