Learn More
SNX27 Rabbit anti-Human, Mouse, DyLight™ 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP145282UV
Catalog No. NBP145282UV
Add to cart
Description
Description
SNX27 Polyclonal antibody specifically detects SNX27 in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
SNX27
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
SNX27
Synthetic peptide made to an internal portion of human SNX27 (within residues 150-200). [Swiss-ProtNo. Q96L92]
0.1 mL
Primary
81609
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
KIAA0488MGC126873, methamphetamine-responsive transcript 1, MGC126871, MGC20471, MRT1, MY014, sorting nexin family member 27, sorting nexin-27
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human, Mouse