Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX29 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen SNX29
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123388
View Documents
Novus Biologicals
NBP30924325UL
25 μg
Each for $106.19
Add to cart
 
NB123387
View Documents
Novus Biologicals
NBP309243100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

SNX29 Polyclonal antibody specifically detects SNX29 in Human samples. It is validated for Western Blot
Specifications

Specifications

SNX29
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
92017
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
A-388D4.1, FLJ00143, FLJ12363, FLJ40609, RUN domain containing 2A, sorting nexin 29
The immunogen is a synthetic peptide directed towards the N-terminal region of Human SNX29. Peptide sequence AVLQHGLKRSRGLALTAAAIKQAAGFASKTETEPVFWYYVKEVLNKHELQ
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit