Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX30 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550772

Catalog No. PA550772


Add to cart

Description

Description

The antibody detects endogenous levels of total SNX30 protein.

SNX9, SNX18 and SNX30 constitute a separate subfamily of PX-BAR-containing sorting nexin (SNX) proteins. May be involved in several stages of intracellular trafficking.
Specifications

Specifications

SNX30
Polyclonal
Unconjugated
SNX30
Liquid
SNX30
Synthetic peptide corresponding to a region derived from internal residues of human sorting nexin family member 30
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.2 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q5VWJ9
ATG24A
Rabbit
IgG
100 μL
-20°C
Primary
401548
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit