Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550769

Catalog No. PA550769


Add to cart

Description

Description

The antibody detects endogenous levels of total SNX5 protein.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein binds to fanconi anemia complementation group A protein, but its function is unknown. This gene results in two transcript variants encoding the same protein.
Specifications

Specifications

SNX5
Polyclonal
Unconjugated
SNX5
Liquid
SNX5
Synthetic peptide corresponding to a region derived from internal residues of human sorting nexin 5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin)
1.6 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9Y5X3, Q9D8U8, B1H267
FLJ10931, SNX5, sorting nexin 5, Sorting nexin-5
Rabbit
IgG
100 μL
-20°C
Primary
27131, 296199, 69178
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit