Learn More
SNX8 Mouse anti-Human, Rat, Clone: 3B6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524455
Catalog No. PIMA524455
Add to cart
Description
Description
Peptide Sequence: MTGRAMDPLP AAAVGAAAEA EADEEADPPA SDLPTPQAIE PQAIVQQVPA PSRMQMPQGN PLLLSHTLQE LLARDTVQVE LIPEKKGLFL KHVEYEVSSQ RFKSSVYRRY NDFVVFQEML LHKFPYRMVP ALPPKRMLGA DREFIEARRR ALKRFVNLVA RHPLFSEDVV LKLFLSFSGS DVQNKLKESA QCVGDEFLNC KLATRAKDFL PADIQAQFAI SRELIRNIYN SFHKLRDRAE RIASRAIDNA ADLLIFGKEL SAIGSDTTPL PSWAALNSST WGSLKQALKG LSVEFALLAD KAAQQGKQEE NDVVEKLNLF LDLLQSYKDL CERHEKGVLH KHQRALHKYS LMKRQMMSAT AQNREPESVE QLESRIVEQE NAIQTMELRN YFSLYCLHQE TQLIHVYLPL TSHILRAFVN SQIQGHKEMS KVWNDLRPKL SCLFAGPHST LTPPCSPPED GLCPH
Sorting nexin (SNX) proteins are members of a large family of hydrophilic PX (phospholipid-binding motif) domain-containing proteins that interact with a variety of receptor types. SNXs are widely expressed, although the tissue distribution of each SNX mRNA varies. The ability of SNXs to bind specific phospholipids, as well as their tendency to form protein-protein complexes, suggests a role for these proteins in cellular membrane trafficking and protein sorting. SNXs may also function specifically in pro-degradative sorting, internalization, endosomal recycling or simply in endosomal sorting. SNXs partially associate with cellular membranes, despite their hydrophilic nature. SNX8 is an ortholog of a yeast protein.
Specifications
Specifications
SNX8
Monoclonal
Unconjugated
SNX8
Liquid
SNX8
SNX8 (AAH21565, 1 a.a. ∽ 465 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human, Rat
ELISA, Western Blot
3B6
PBS with no preservative; pH 7.4
Q9Y5X2
Mvp1
Mouse
IgG2b, kappa
100 μg
-20° C
Primary
288504, 29886