Learn More
SNX9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 157211AP
Catalog No. 50-173-0431
Add to cart
Description
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but does contain a SH3 domain near its N-terminus. This protein interacts with the cytoplasmic domains of the precursor but not the processed forms of a disintegrin and metalloprotease domain 9 and 15. This protein binds the beta-appendage domain of adaptor protein 2 and may function to assist adaptor protein 2 in its role at the plasma membrane. This protein interacts with activated Cdc42-associated kinase-2 to regulate the degradation of epidermal growth factor receptor protein.
Specifications
Specifications
SNX9
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91VH2, Q9Y5X1
MST155, MSTP155, Protein SDP1, SDP1, SH3PX1, SH3PXD3A, SNX9, sorting nexin 9, WISP
Rabbit
IgG
150 μL
-20°C
Primary
51429, 66616, 683687
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
SNX9
Liquid
SNX9
SNX9 Fusion Protein Ag8440
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat