Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX9 Monoclonal Antibody (OTI2F1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA525154
Description
SNX9 Monoclonal antibody specifically detects SNX9 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
SNX9 | |
Monoclonal | |
0.78 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y5X1 | |
Snx9 | |
Full length human recombinant protein of SNX9 produced in HEK293T cell | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2F1 | |
Unconjugated | |
Snx9 | |
2700073N08Rik; MST155; MSTP155; OTTHUMP00000040083; Protein SDP1; RP11-266C7.10; SDP1; SH3 and PX domain-containing protein 1; SH3 and PX domain-containing protein 3A; SH3PX1; SH3PXD3A; Snx9; sorting nexin 9; sorting nexin-9; Wiskott-Aldrich syndrome protein (WASP) interactor protein; WISP | |
Mouse | |
Affinity Chromatography | |
RUO | |
51429 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction