Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Soap solution, According to Boutron and Boudet, DIN 8106, Honeywell Fluka™
According to Boutron and Boudet, DIN 8106
Supplier: Honeywell Chemicals 353661L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Soap solution | |
MFCD00004595 | |
22°C (71.6°F) | |
CO | |
32.04 |
CH4O | |
1 L | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol |
Safety and Handling
P210-P280-P305 + P351 + P338-P337 + P313-P403 + P235
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title