Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOAT2 Mouse anti-Human, Clone: OTI2D3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526484

Catalog No. PIMA526484


Add to cart

Description

Description

This gene is a member of a small family of acyl coenzyme A:cholesterol acyltransferases. The gene encodes a membrane-bound enzyme localized in the endoplasmic reticulum that produces intracellular cholesterol esters from long-chain fatty acyl CoA and cholesterol. The cholesterol esters are then stored as cytoplasmic lipid droplets inside the cell. The enzyme is implicated in cholesterol absorption in the intestine and in the assembly and secretion of apolipoprotein B-containing lipoproteins such as very low density lipoprotein (VLDL). Several alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known.
Specifications

Specifications

SOAT2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O75908
SOAT2, ACACT2, ACAT2, ARGP2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8435
Immunohistochemistry (Paraffin)
OTI2D3
Unconjugated
SOAT2
Liquid
SOAT2
Human recombit protein fragment corresponding to amino acids 1-120 of SOAT2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit