Learn More
DWK Life Sciences Socorex™ Qualitix™ 10 μL Micropipette Tips
Certified free of nuclease, endotoxin and ATP
Manufacturer:  DWK Life Sciences 3090.0010FRS
Catalog No. 12-141-505
Add to cart
Description
Description
  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
Specifications
Specifications
10 µL
Non-sterile
Autoclavable
RNase, DNase, and pyrogen free
12 x 96/Rack
Non-Filtered
Natural
31.4 mm
Rack
Micropipette tips