Learn More
DWK Life Sciences Socorex™ Qualitix™ 10 μL Micropipette Tips
Certified free of nuclease, endotoxin and ATP
$52.25 - $135.00
Specifications
Volume (Metric) 10 µL
Color Natural
Autoclavable Autoclavable
Length (Metric) 31.4 mm
Clean Claims RNase, DNase, and pyrogen free
View More Specs
Products 2
Catalog Number Mfr. No. Filtered Format Sterility Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Filtered Format Sterility Packaging Price Quantity & Availability  
12-141-503
View Documents
DWK Life Sciences
3090.0010B
Non-Filtered Bagged Non-sterile 1000/Bag
Case of 1000 for $52.25
Add to cart
12-141-504
View Documents
DWK Life Sciences
3090.0010R
Filtered Rack Sterile 12 x 96/Rack
Case of 1152 for $135.00
Add to cart
Description
Description
  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
Specifications
Specifications
10 µL
Autoclavable
RNase, DNase, and pyrogen free
Natural
31.4 mm
Micropipette tips