Learn More
DWK Life Sciences Socorex™ Qualitix™ 10 μL Micropipette Tips Low Binding
Certified free of nuclease, endotoxin and ATP
$147.00 - $169.00
Specifications
Volume (Metric) 10 µL
Color Natural
Autoclavable Autoclavable
Length (Metric) 31.4 mm
Clean Claims RNase, DNase, and pyrogen free
View More Specs
Products 2
Catalog Number Mfr. No. Filtered Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Filtered Sterility Price Quantity & Availability  
12-141-524
View Documents
DWK Life Sciences
3090.0010PR
Filtered Non-sterile
Case of 1152 for $147.00
Add to cart
12-141-525
View Documents
DWK Life Sciences
3090.0010PRS
Non-Filtered Sterile
Case of 1152 for $169.00
Add to cart
Description
Description
  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
Specifications
Specifications
10 µL
Autoclavable
RNase, DNase, and pyrogen free
12 x 96/Rack
Natural
31.4 mm
Rack
Micropipette tips