Learn More
DWK Life Sciences Socorex™ Qualitix™ 1000 μL Micropipette Tips
Certified free of nuclease, endotoxin and ATP
$44.37 - $158.45
Specifications
Clean Claims RNase, DNase, and pyrogen free
Autoclavable Autoclavable
Color Natural
Volume (Metric) 1000 µL
Length (Metric) 88.9 mm
View More Specs
Products 3
Catalog Number Mfr. No. Filtered Format Sterility Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Filtered Format Sterility Packaging Price Quantity & Availability  
12-141-519
View Documents
DWK Life Sciences
3070.1000B
Non-Filtered Bagged Non-sterile 1000 Pk.
Case of 1000 for $44.37
Add to cart
12-141-520
View Documents
DWK Life Sciences
3070.1000R
Non-Filtered Rack Non-sterile 6 x 96/Rack
Case of 576 for $82.39
Add to cart
12-141-521
View Documents
DWK Life Sciences
3070.1000FRS
Filtered Rack Sterile 6 x 96/Rack
Case of 576 for $158.45
Add to cart
Description
Description
  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
Specifications
Specifications
RNase, DNase, and pyrogen free
Natural
88.9 mm
Autoclavable
1000 µL
Micropipette tips