Learn More
DWK Life Sciences Socorex™ Qualitix™ 200 μL Micropipette Tips Low Binding
Certified free of nuclease, endotoxin and ATP
$116.00 - $131.00
Specifications
Volume (Metric) 200 µL
Color Natural
Autoclavable Autoclavable
Length (Metric) 49.6 mm
Clean Claims RNase, DNase, and pyrogen free
View More Specs
Products 2
Catalog Number Mfr. No. Filtered Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Filtered Sterility Price Quantity & Availability  
12-141-526
View Documents
DWK Life Sciences
3070.0200PR
Non-Filtered Non-sterile
Case of 960 for $116.00
Add to cart
12-141-527
View Documents
DWK Life Sciences
3070.0200PRS
Filtered Sterile
Case of 960 for $131.00
Add to cart
Description
Description
  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
Specifications
Specifications
200 µL
Autoclavable
RNase, DNase, and pyrogen free
10 x 96/Rack
Natural
49.6 mm
Rack
Micropipette tips