Learn More
DWK Life Sciences Socorex™ Qualitix™ 350 μL Micropipette Tips
Certified free of nuclease, endotoxin and ATP
Specifications
Volume (Metric) 350 µL
Color Natural
Autoclavable Autoclavable
Length (Metric) 58.8 mm
Clean Claims RNase, DNase, and pyrogen free
View More Specs
Products 2
Catalog Number Mfr. No. Format Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Format Packaging Price Quantity & Availability  
12-141-516
View Documents
DWK Life Sciences
308.0350B
Bagged 1000/Bag
Case of 1000 for $95.00
N/A
12-141-517
View Documents
DWK Life Sciences
308.0350R
Rack 10 x 96/Rack
Case of 960 for $183.00
N/A
Description
Description
  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
Specifications
Specifications
350 µL
Autoclavable
RNase, DNase, and pyrogen free
Natural
58.8 mm
Micropipette tips