Learn More
DWK Life Sciences Socorex™ Qualitix™ Micropipette Tips, Gel Loading
Certified free of nuclease, endotoxin and ATP
$119.27 - $131.37
Specifications
Clean Claims RNase, DNase, and pyrogen free
Autoclavable Autoclavable
Color Natural
Format Rack
Product Type Micropipette tips, Gel loading
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Length (Metric) Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Length (Metric) Packaging Price Quantity & Availability  
12-141-507
View Documents
DWK Life Sciences
302.0010GR
10 µL 71 mm 4 x 204/Rack
Case of 816 for $131.37
Add to cart
12-141-515
View Documents
DWK Life Sciences
3080.0200GR
200 µL 82.5 mm 6 x 96/Rack
Case of 576 for $119.27
Add to cart
Description
Description
  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
Specifications
Specifications
RNase, DNase, and pyrogen free
Natural
Micropipette tips, Gel loading
Autoclavable
Rack