Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOCS3 Mouse anti-Human, Clone: OTI11C5, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502959
Description
Accumulating evidence demonstrates that cytokine receptor signaling is negatively regulated by a family of Src homology 2 domain-containing adaptor molecules Termed SOCS (Suppressor of Cytokine Signaling). To date, there are eight members of SOCS family have been recognized, they are SOCS-1, 2, 3, 4, 5, 6, 7 and CIS. Structurally the SOCS proteins are composed of an N-terminal region of variable length and amino acid composition, a central SH2 domain, and a previously unrecognized C-terminal motif that we have called the SOCS box. The SOCS proteins appear to form part of a classical negative feed back loop that regulates cytokine signal transduction via a STAT-induced transcriptional mechanism. Transcription of each of the SOCS genes occurs rapidly in vitro and in vivo in response to cytokines, and once produced, the various members of the SOCS family appear to inhibit signaling in different ways.Specifications
SOCS3 | |
Monoclonal | |
Unconjugated | |
SOCS3 | |
Liquid | |
SOCS3 | |
Full length human recombit protein of human SOCS3 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI11C5 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O14543 | |
ATOD4, CIS3, Cish3, SOCS-3, SSI-3, SSI3 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
9021 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title