Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOCS4 Mouse anti-Human, Clone: OTI5H9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527409

Catalog No. PIMA527409


Add to cart

Description

Description

The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

SOCS4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WXH5
SOCS7, SH2 domain containing SOCS box protein, SOCS-4, SOCS-7, suppressor of cytokine signaling 7
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
122809
Western Blot
OTI5H9
Unconjugated
SOCS4
Liquid
SOCS4
Full length human recombit protein of SOCS4 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit