Learn More
BioVision IncSupplier Diversity Partner Superoxide Dismutase (SOD) Activity Colorimetric Assay Kit: Superoxide Dismutase (SOD) Activity Assay Kit: Measure SOD Activity in Tissues, Cells, Blood etc. within 30 min. Simple, Convenient & Accurate. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K335100
Superoxide dismutase (SOD) is one of the most important antioxidative enzymes. It catalyzes the dismutation of the superoxide anion into hydrogen peroxide and molecular oxygen. The sensitive SOD assay kit utilizes WST-1 that produces a water-soluble formazan dye upon reduction with superoxide anion. The rate of the reduction with a superoxide anion is linearly related to the xanthine oxidase (XO) activity, and is inhibited by SOD (below). Therefore, the inhibition activity of SOD can be determined by a colorimetric method. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9701