Learn More
Novus Biologicals™ SOD (Superoxide Dismutase) Activity Assay Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Quantity | 96 Tests |
| Research Category | Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors |
| Species | Non-species specific |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.