Learn More
SOD1 Mouse anti-Bovine, Human, Clone: 2F5, Abnova™
Mouse monoclonal antibody raised against bovine SOD1.
Manufacturer:  Abnova Corporation MAB12978
Catalog No. 89-934-944
Add to cart
Description
Description
Specifications
Specifications
SOD1
2F5
Mouse monoclonal antibody raised against bovine SOD1.
In PBS (0.1% BSA, 0.09% sodium azide).
SOD1L1
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Bovine, Human
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Unconjugated
ELISA Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
SOD1
Bovine erythrocytes.
Protein G purification
RUO
Primary
281495