Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD3/EC-SOD Mouse anti-Guinea Pig, Human, Mouse, Rat, Clone: 4GG11G6, Novus Biologicals™

Mouse MonoclonalAntibody

$235.00 - $358.00

Specifications

Antigen SOD3/EC-SOD
Clone 4GG11G6
Dilution Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
NB59688100U View Documents Novus BiologicalsSupplier Diversity Partner
NBP259688100UG
100 ug Unconjugated Each for $358.00
Add to cart
 
NB5968825UG View Documents Novus BiologicalsSupplier Diversity Partner
NBP25968825UG
25 ug Unconjugated Each for $235.00
Add to cart
 
Description

Description

SOD3/EC-SOD Monoclonal antibody specifically detects SOD3/EC-SOD in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

SOD3/EC-SOD
Western Blot 1:1000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100
Purified
Human, Mouse, Rat, Guinea Pig
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Human extracellular SOD purified from aortas
Monoclonal
Protein G purified
Detects extracellular SOD ~35kDa.
4GG11G6
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Mouse
RUO
6649
IgG1 Kappa
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit