Soda lime, indicating, ACS

Manufacturer: Alfa Aesar 44697A4

Quantity 2.5kg

 View more versions of this product

Catalog No. AA44697A4

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 8006-28-8
    MDL Number MFCD00134094
    UN Number UN1907
    Merck Index 14,8569
    Chemical Name or Material Soda lime, indicating, For ACS analysis; Variable mix of sodium hydroxide with calcium oxide or hydroxide
    Grade ACS Grade

    Specifications

    Physical Form 4 To 8 Mesh
    Quantity 2.5kg
    UN Number UN1907
    TSCA TSCA
    Merck Index 14,8569
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    I
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: SODA LIME
    CAS 8006-28-8
    MDL Number MFCD00134094
    Chemical Name or Material Soda lime, indicating, For ACS analysis; Variable mix of sodium hydroxide with calcium oxide or hydroxide
    Grade ACS Grade

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    I

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit