Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soda Lime, Indicating Type, 4 to 8 Mesh, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

8006-28-8
CaHNaO2
96.074
HUAUNKAZQWMVFY-UHFFFAOYSA-M
66545795

High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Mesh Size 4 to 8 Mesh
Packaging Poly Bottle
SMILES [OH-].O=[Ca].[Na+]
IUPAC Name sodium;oxocalcium;hydroxide
Chemical Name or Material Soda Lime, Indicating Type
Grade Baker Analyzed™ ACS Reagent