Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soda Lime, Indicating Type, 4 to 8 Mesh, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

8006-28-8
CaHNaO2
96.074
HUAUNKAZQWMVFY-UHFFFAOYSA-M
66545795

High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
CAS 8006-28-8
Mesh Size 4 to 8 Mesh
Packaging Poly Bottle
Molecular Formula CaHNaO2
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES [OH-].O=[Ca].[Na+]
IUPAC Name sodium;oxocalcium;hydroxide
Molecular Weight (g/mol) 96.074
PubChem CID 66545795
Chemical Name or Material Soda Lime, Indicating Type
Grade Baker Analyzed™ ACS Reagent
Show More Show Less