Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
SODD/BAG4 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF4350SP
Catalog No. AF4350SP
This item is not returnable. View return policy
Description
Description
SODD/BAG4 Polyclonal antibody AF4350SP specifically detects SODD/BAG4 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
SODD/BAG4
LYOPH
Western Blot 0.5 μg/mL, Immunocytochemistry 5-15 μg/mL, Knockout Validated
O95429
BAG4, BAG-4, BAG-family molecular chaperone regulator-4, Bcl-2-associated athanogene 4, BCL2-associated athanogene 4, Silencer of death domains, SODDBAG family molecular chaperone regulator 4
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human SODD/BAG4 in Western blots.
Human
Immunoassay, Immunocytochemistry, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
BAG4
E. coli-derived recombinant human SODD/BAG4 Glu122-Leu457 Accession # O95429
Antigen Affinity-purified
RUO
Primary
9530
Reconstitute at 0.2 mg/mL in sterile PBS.