Alfa Aesar™ Sodium 1-hexanesulfonate monohydrate, 99%

Manufacturer: Alfa Aesar™ 3219018

Quantity 50g

 View more versions of this product

Catalog No. AA3219018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 207300-91-2
    Linear Formula CH3(CH2)5SO3Na·H2O
    Formula Weight 206.24 (188.22 Anhydrous)
    MDL Number MFCD00149549
    Beilstein 3727014
    Synonym 1-Hexanesulfonic acid sodium salt monohydrate
    Chemical Name or Material Sodium 1-Hexanesulfonate Monohydrate

    Specifications

    Formula Weight 206.24 (188.22 Anhydrous)
    Physical Form Crystalline
    MDL Number MFCD00149549
    Synonym 1-Hexanesulfonic acid sodium salt monohydrate
    Quantity 50g
    TSCA TSCA
    Linear Formula CH3(CH2)5SO3Na·H2O
    CAS 207300-91-2
    Beilstein 3727014
    Color White
    Chemical Name or Material Sodium 1-Hexanesulfonate Monohydrate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit