Alfa Aesar™ Sodium 1-octanesulfonate monohydrate, 99+%

Manufacturer: Alfa Aesar™ 1576418

Quantity 50g

 View more versions of this product

Catalog No. AA1576418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 207596-29-0
    Formula Weight 234.29 (216.28 Anhydrous)
    MDL Number MFCD00007544
    Synonym 1-Octanesulfonic acid sodium salt monohydrate
    Chemical Name or Material Sodium 1-Octanesulfonate Monohydrate

    Specifications

    Formula Weight 234.29 (216.28 Anhydrous)
    Physical Form Crystalline
    MDL Number MFCD00007544
    CAS 207596-29-0
    Synonym 1-Octanesulfonic acid sodium salt monohydrate
    TSCA TSCA
    Melting Point >300°C
    Quantity 50g
    Chemical Name or Material Sodium 1-Octanesulfonate Monohydrate
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit