Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium 1-pentyl sulfate, 99%

Click to view available options
Quantity:
5g
25g
556-76-3
C5H11NaO4S
190.189
MFCD00041882
KLZYTGLGWPHIDV-UHFFFAOYSA-M
sodium pentyl sulfate,pentyl sodium sulphate,sodium 1-pentyl sulfate,pentyl sodium sulfate,sulfuric acid, monopentyl ester, sodium salt,sodium amyl sulfate,naxolate as-lg95,sulfuric acid, monopentyl ester, sodium salt 1:1,sodium pentyl sulphate,n-amyl sulfate sodium salt
23680281
sodium;pentyl sulfate
CCCCCOS(=O)(=O)[O-].[Na+]
556-76-3
C5H11NaO4S
190.189
MFCD00041882
KLZYTGLGWPHIDV-UHFFFAOYSA-M
sodium pentyl sulfate,pentyl sodium sulphate,sodium 1-pentyl sulfate,pentyl sodium sulfate,sulfuric acid, monopentyl ester, sodium salt,sodium amyl sulfate,naxolate as-lg95,sulfuric acid, monopentyl ester, sodium salt 1:1,sodium pentyl sulphate,n-amyl sulfate sodium salt
23680281
sodium;pentyl sulfate
CCCCCOS(=O)(=O)[O-].[Na+]
Applications
Sodium 1-pentyl sulfate is used as a pharmaceutical intermediate, in chemical research.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Quantity 25g
Beilstein 4163608
Solubility Information Soluble in water.
Formula Weight 190.2
Percent Purity 99%
Chemical Name or Material Sodium 1-pentyl sulfate

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-140-9

TSCA : Yes

Recommended Storage : Ambient temperatures