Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium 1-propanethiolate, 85%

Click to view available options
Quantity:
5g
25g
6898-84-6
C3H7NaS
98.139
MFCD00043318
XVSFHIIADLZQJP-UHFFFAOYSA-M
sodium 1-propanethiolate,sodium propanethiolate,1-propanethiol sodium salt,sodium propane-1-thiolate,1-propanethiol, sodium salt,sodium propylsulfanide,propylsulfanylsodium,sodium thiopropoxide,1-propanethiol, sodium salt 1:1,sodium propyl sulfide
4681725
sodium;propane-1-thiolate
CCC[S-].[Na+]
6898-84-6
C3H7NaS
98.139
MFCD00043318
XVSFHIIADLZQJP-UHFFFAOYSA-M
sodium 1-propanethiolate,sodium propanethiolate,1-propanethiol sodium salt,sodium propane-1-thiolate,1-propanethiol, sodium salt,sodium propylsulfanide,propylsulfanylsodium,sodium thiopropoxide,1-propanethiol, sodium salt 1:1,sodium propyl sulfide
4681725
sodium;propane-1-thiolate
CCC[S-].[Na+]
Odor Stench
Sensitivity Air & Moisture Sensitive
Color Gray to White
Quantity 25g
Linear Formula CH3CH2CH2SNa
UN Number UN3263
Beilstein 3556024
Formula Weight 98.14
Physical Form Solid
Percent Purity 85%
Chemical Name or Material Sodium 1-propanethiolate
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.

TSCA : TSCA