Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate

Click to view available options
Quantity:
1g
5g
25g
1191-50-0
C14H29NaO4S
316.432
MFCD00007468
UPUIQOIQVMNQAP-UHFFFAOYSA-M
sodium tetradecyl sulfate,sodium myristyl sulfate,niaproof 4,sodium tetradecyl sulphate,tesapon k 14,texapon k 14,trombavar,natri tetradecylsulfas,sotradecol sodium,tetradecyl sodium sulfate
23665770
sodium;tetradecyl sulfate
CCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
1191-50-0
C14H29NaO4S
316.432
MFCD00007468
UPUIQOIQVMNQAP-UHFFFAOYSA-M
sodium tetradecyl sulfate,sodium myristyl sulfate,niaproof 4,sodium tetradecyl sulphate,tesapon k 14,texapon k 14,trombavar,natri tetradecylsulfas,sotradecol sodium,tetradecyl sodium sulfate
23665770
sodium;tetradecyl sulfate
CCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Applications
Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Quantity 5g
Melting Point 182°C to 192°C
Beilstein 3578122
Formula Weight 316.44
Percent Purity 95%
Chemical Name or Material Sodium 1-tetradecyl sulfate
Assay cont. up to ≈5% sodium methyl sulfate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XB8660000

TSCA : TSCA